missing translation for 'onlineSavingsMsg'
Learn More

BINDER™ Viewing Window and Interior Lighting

Product Code. 16494139 Shop All BINDER Products
Click to view available options
For Use With (Equipment):
BINDER Chambers ED 260
BINDER Chambers ED 56
BINDER Chambers ED 720
BINDER Chambers FD 23
BINDER Chambers FD 260
BINDER Chambers FD 56
BINDER Chambers FD 720
BINDER Chambers FED 260
BINDER Chambers FED 400
BINDER Chambers FED 56
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 16494139

Brand: BINDER™ 80121259

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

This Viewing Window and Interior Lighting is designed for use with assorted BINDER™ Drying and Heating Chambers. Insulated viewing window and interior lighting allows observation without interfering with internal conditions.

  • Viewing window with triple insulating glass in door(s)
  • 15 or 30 W interior lighting, depending on the model

Specifications

Description Viewing window and interior lighting - 230 V option model, viewing window in door, 180 x 180 mm, and 15 W interior lighting
For Use With (Equipment) BINDER Chambers FD 23
Type Viewing Window with Interior Lighting
Insulation Triple Insulating Glass
Width (Metric) 180 mm
Height (Metric) 180 mm
Wattage 15 W
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.