Learn More
Syngene™ Viewing Platform for Gbox Systems
GREENER_CHOICE
Product Code. 12836974 Shop All Syngene Products
Change view
Click to view available options
For Use With (Equipment):
Syngene Gbox
Unit Size:
Each
Product Code. For Use With (Equipment) unitSize
12836974 Syngene Gbox Each
1 options
Product Code. 12836974 Supplier Syngene™ Supplier No. GXVP
Only null left
Add to basket
Add to basket
Viewing Platform for Gbox Systems.
  • Viewing platform for GBOX system
  • To be used in place of transilluminator
Specifications
Product Line Syngene
For Use With (Application) Viewing platform
For Use With (Equipment) Syngene Gbox
Voltage 100/115/220/240 V
Product Type Viewing Platform