Learn More
IGFBP-2 Antibody (CL13563) [Biotin], Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP3-24568B
Product Code. 30287259
  • £558.00 / 0.10mL
Description
IGFBP-2 Monoclonal antibody specifically detects IGFBP-2 in Human samples. It is validated for Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY
Specifications
Specifications
IGFBP-2
Monoclonal
Biotin
BP2, IBP2, IBP-2, IGF-binding protein 2, IGFBP-2, IGF-BP53, insulin-like growth factor binding protein 2 (36kD), insulin-like growth factor binding protein 2, 36kDa, insulin-like growth factor-binding protein 2
This antibody was developed using a synthetic peptide derived from P18065, with the exact immunogen sequence remaining proprietary.
0.1 mL
Cancer, Cell Biology, Cytokines and Growth Factors, Signal Transduction
3485
Store at 4⪚C in the dark.
IgG1
Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin)
CL13563
PBS
Mouse
Protein A purified