Learn More
Invitrogen™ eIF4G Recombinant Rabbit Monoclonal Antibody (0B8X7)
Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA557937
This item is not returnable.
View return policy
Description
Immunogen sequence: GSNPGPESEG SGVPPRPEEA DETWDSKEDK IHNAENIQPG EQKYEYKSDQ WKPLNLEEKK RYDREFLLGF QFIFASMQKP EGLPHISDVV LDKANKTPLR.
The protein encoded by this gene is a component of the protein complex EIF4F, which is involved in the recognition of the mRNA cap, ATP-dependent unwinding of 5'-terminal secondary structure, and recruitment of mRNA to the ribosome. Alternative splicing results in five transcript variants encoding four distinct isoforms.
Specifications
| eIF4G | |
| Recombinant Monoclonal | |
| 1.1 mg/mL | |
| PBS with 0.05% BSA, 50% glycerol and 0.05% ProClin 300; pH 7.3 | |
| Q6NZJ6 | |
| EIF4G1 | |
| A synthetic peptide corresponding to a sequence within amino acids 551-650 of human EIF4G1 (NP_937884.2). | |
| 20 μL | |
| Primary | |
| Mouse, Rat | |
| Antibody | |
| IgG |
| ELISA, Immunohistochemistry (Paraffin), Western Blot | |
| 0B8X7 | |
| Unconjugated | |
| EIF4G1 | |
| DKFZp686A1451; E030015G23Rik; EIF4F; EIF4G; eIF-4G 1; EIF4G1; eIF-4G1; EIF4-gamma; eIF-4-gamma 1; EIF4GI; eIF4GI protein; eucaryotic translation initiation factor 4G; Eukaryotic translation initiation factor 4 gamma 1; eukaryotic translation initiation factor 4 gamma, 1; eukaryotic translation initiation factor 4, gamma 1; OTTHUMP00000210577; OTTHUMP00000210586; OTTHUMP00000210587; OTTHUMP00000210588; OTTHUMP00000210595; OTTHUMP00000210602; OTTHUMP00000210603; P220; PARK18 | |
| Rabbit | |
| Protein A | |
| RUO | |
| 208643, 287986 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |