Learn More
Invitrogen™ Gevokizumab Recombinant Monoclonal Antibody
Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA559373
This item is not returnable.
View return policy
Description
For reconstitution, add sterile, distilled water to achieve a final antibody concentration of 1 mg/mL. Gently shake to solubilize the protein completely. Do not vortex. Reconstituted products should be stored at -80 °.
Gevokizumab has been used in trials studying the treatment of Acne Vulgaris, Osteoarthritis, Behcet's Uveitis, Pyoderma Gangrenosum, and Behcet's Disease Uveitis, among others. Gevokizumab acts as a modulator of cytokine imbalance in IL-1 mediated disease states. It has a very high binding affinity of 300fM and blocks the activation of IL-1 receptors.
Specifications
| Gevokizumab | |
| Recombinant Monoclonal | |
| Unconjugated | |
| XOMA 052 | |
| Protein A | |
| RUO | |
| Human | |
| Antibody | |
| IgG2SA |
| ELISA, Flow Cytometry, Functional Assay, Surface Plasmon Resonance | |
| 1 mg/mL | |
| 25mM histidine with 8% sucrose, 0.01% Tween 80 and no preservative; pH 6.2 | |
| Human | |
| 100 μg | |
| Primary | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |