Learn More
Novus Biologicals™ Human RelA/NFkB p65 - Small Sample ELISA Kit (Colorimetric)
Product Code. 30279447 Shop All Bio Techne Products
Click to view available options
Quantity:
1 Kit
Unit Size:
Each
Product Code. 30279447
Brand: Novus Biologicals™ NBP343214
Colorimetric ELISA Kit
TRUSTED_SUSTAINABILITY
Specifications
Assay Range 0.156 to 10 ng/mL
Assay Sensitivity 0.058 ng/mL
Detection Method Colorimetric
Immunoassay Kit Format Pre-coated 96T Strip Plate
Sample Type Tissue Homogenates, Cell Lysates, Other Biological Fluids
Target Species Human
For Use With (Application) ELISA
Gene ID (Entrez) 5970
Gene Symbol RELA
Kit Contents Pre-coated 96T Strip Plate, Plate sealer for 96 wells, Standard, Standard Diluent, Detection Reagent A, Assay Diluent A, Detection Reagent B, Assay Diluent B, TMB Substrate, Stop Solution, Wash Buffer (30X concentrate), Instruction manual
Quantity 1 Kit
Research Discipline Apoptosis, Cancer, Cell Biology, Immune System Diseases, Immunology, Innate Immunity, Neurodegeneration, Neuroscience, Phospho Specific, Signal Transduction, Transcription Factors and Regulators
Storage Requirements Storage of components varies. See protocol for specific instructions.
Synonym MGC131774, nf kb p65, NFkB p65, NF-kB p65, NFKB3v-rel avian reticuloendotheliosis viral oncogene homolog A (nuclear factor ofkappa light polypeptide gene enhancer in B-cells 3 (p65)), Nuclear factor NF-kappa-B p65 subunit, Nuclear factor of kappa light polypeptide gene enhancer in B-cells 3transcription factor p65, p65, RelA, rela p65, v-rel reticuloendotheliosis viral oncogene homolog A (avian), v-rel reticuloendotheliosis viral oncogene homolog A, nuclear factor of kappalight polypeptide gene enhancer in B-cells 3, p65