Learn More
Pregnenolone Antibody (D1-2), Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP3-43428
Product Code. 30279413
  • £252.00 / 100µL
Description
Pregnenolone Monoclonal antibody specifically detects Pregnenolone in Nonspecies specific samples. It is validated for Immunohistochemistry,Immunoprecipitation,Western Blot,Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Pregnenolone
Monoclonal
1 mg/mL
Western Blot 0.01-2ug/mL, Immunohistochemistry 5-20 μg/mL, Immunocytochemistry/ Immunofluorescence 5-20 μg/mL, Immunoprecipitation
Mouse
Protein A or G purified
RUO
Non-species specific
Purified
Immunohistochemistry, Immunoprecipitation, Western Blot, Immunocytochemistry/Immunofluorescence
D1-2
Unconjugated
0.01 M PBS, pH 7.4, 50% glycerol
OVA - Pregnenolone
100 μL
Primary
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Not Available