Learn More
Invitrogen™ COBRA1 Recombinant Rabbit Monoclonal Antibody (24GB3740)
Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA557112
This item is not returnable.
View return policy
Description
COBRA1 Recombinant Monoclonal Antibody for Western Blot, ICC/IF
NELFB is a subunit of negative elongation factor (NELF), which also includes NELFA (WHSC2), either NELFC or NELFD (TH1L), and NELFE (RDBP), a heterodimer of SPT4 (SUPT4H1), to cause transcriptional pausing of RNA polymerase II (Narita et al., 2003).
Specifications
| COBRA1 | |
| Recombinant Monoclonal | |
| Unconjugated | |
| NELFB | |
| A730008L03Rik; AB041607; AI663983; Cobra1; cofactor of BRCA1; DKFZp586B0519; KIAA1182; MNCb-5210; Negative elongation factor B; negative elongation factor complex member B; negative elongation factor complex member B, Cobra1; negative elongation factor protein B; NELFB; NELF-B; ortholog of human cofactor of BRCA1 (COBRA1); RGD1307832; RP13-122B23.3 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 25920, 311796, 58202 | |
| -20°C | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 24GB3740 | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q8C4Y3, Q8WX92 | |
| NELFB | |
| A synthesized peptide derived from human NELFB (aa 1-200). | |
| 20 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |