Learn More
Invitrogen™ PRMT6 Recombinant Rabbit Monoclonal Antibody (24GB1585)
Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA557631
This item is not returnable.
View return policy
Description
PRMT6 Recombinant Monoclonal Antibody for Western Blot, Flow
Protein arginine N-methyltransferase 6 (PRMT6) PRMT6 demonstrates type I PRMT activity, capable of forming both omega-N monomethylarginine and asymmetric dimethylarginine derivatives on the recombinant glycine- and arginine-rich substrates. cts as a transcriptional coactivator of a number of steroid hormone receptors including ESR1, ESR2, PGR and NR3C1. Promotes fasting-induced transcriptional activation of the gluconeogenic program through methylation of the CRTC2 transcription coactivator. May play a role in innate immunity against HIV-1 in case of infection by methylating and impairing the function of various HIV-1 proteins such as Tat, Rev and Nucleocapsid protein p7.
Specifications
| PRMT6 | |
| Recombinant Monoclonal | |
| Unconjugated | |
| PRMT6 | |
| AW124876; BB233495; FLJ10559; FLJ51477; heterogeneous nuclear ribonucleoprotein methyltransferase-like protein 6; Histone-arginine N-methyltransferase PRMT6; HMT1 hnRNP methyltransferase-like 6; Hrmt1l6; PRMT6; PRMT-6; protein arginine methyltransferase 6; protein arginine N-methyltransferase 6 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 295384, 55170, 99890 | |
| -20°C | |
| Liquid |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| 24GB1585 | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q6NZB1, Q96LA8 | |
| PRMT6 | |
| A synthesized peptide derived from human PRMT6 (aa 203-226). | |
| 20 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido