Learn More
Invitrogen™ ULK1 Recombinant Rabbit Monoclonal Antibody (24GB2195)
Rabbit Recombinant Monoclonal Antibody
Brand:  Invitrogen™ MA557310
 View more versions of this product
Product Code. 30275510
  • £126.00 / 20µL
Description
ULK1 Recombinant Monoclonal Antibody for Western Blot, ICC/IF, Flow
ULK1 is a serine/threonine protein kinase that plays critical role during initial stages of autophagy which is a vital response to nutrient starvation. The conserved C-terminal domain (CTD) of ULK1 controls the regulatory function and localization of the protein. Knockdown of ULK1 inhibits the autophagic response as well as inhibiting rapamycin-induced autophagy consistent with a role downstream of mTOR. ULK1 forms a complex with FIP200 and ATG13 and this complex is essential for starvation-induced autophagy. Both FIP200 and ATG13 are critical for correct localization of ULK1 to the pre-autophagosome and stability of ULK1 protein. ULK1 is phosphorylated by the mTOR pathway in a nutrient starvation-regulated manner.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
ULK1
Recombinant Monoclonal
0.33 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
O70405, O75385
ULK1
A synthesized peptide derived from human ULK1 (aa 950-1050).
20 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Flow Cytometry, Western Blot, Immunocytochemistry
24GB2195
Unconjugated
ULK1
APG1; ATG 1; ATG1; ATG1 autophagy related 1 homolog; ATG1A; AU041434; Autophagy-related protein 1 homolog; C. elegans; EC:2.7.11.1; FLJ38455; hATG1; KIAA0722; mKIAA0722; Serine/threonine-protein kinase ULK1; serine/threonine-protein kinase Unc51.1; ULK 1; Ulk1; UNC 51; UNC51; unc-51 like autophagy activating kinase 1; unc-51 like kinase 1; Unc51.1; unc-51-like kinase 1
Rabbit
Affinity chromatography
RUO
22241, 360827, 8408
-20°C