Learn More
Invitrogen™ CHX10 Recombinant Rabbit Monoclonal Antibody (24GB2590)
Rabbit Recombinant Monoclonal Antibody
Brand:  Invitrogen™ MA557316
 View more versions of this product
Product Code. 30275269
  • £126.00 / 20µL
Description
CHX10 Recombinant Monoclonal Antibody for Western Blot, ICC/IF, Flow
CHX10 encodes a homeobox protein originally described as a retina-specific transcription factor. Mutations in this gene are associated with microphthalmia, cataracts and iris abnormalities.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
CHX10
Recombinant Monoclonal
3 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
P58304, Q61412
VSX2
A synthesized peptide derived from human CHX10 (aa 330-361).
20 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Flow Cytometry, Western Blot, Immunocytochemistry
24GB2590
Unconjugated
VSX2
C. elegans ceh-10 homeo domain containing homolog; ceh-10 homeo domain containing homolog; ceh-10 homeodomain containing homolog; ceh-10 homeodomain-containing homolog; CHX10; Homeobox protein CHX10; HOX10; Hox-10; MCOP2; MCOPCB3; ocular retardation; or; RET1; Visual system homeobox 2; Vsx2
Rabbit
Affinity chromatography
RUO
12677, 171360, 338917
-20°C