Learn More
Invitrogen™ Human ZNF341 (aa 373-445) Control Fragment Recombinant Protein
Product Code. 30213003
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
Product Code. 30213003
Brand: Invitrogen™ RP105416
Recombinant Protein
Highest antigen sequence indentity to the following orthologs: Mouse (84%), Rat (84%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-84745 (PA5-84745. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.
The ZNF341 gene is located on chromosome 20.
TRUSTED_SUSTAINABILITY
Specifications
Accession Number Q9BYN7
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 84905
Name Human ZNF341 (aa 373-445) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias dJ553F4.3; ZFP341; zinc finger protein 341; ZNF341
Common Name ZNF341
Gene Symbol ZNF341
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence KVWPPGHSGGTVSRNSVTVQVMALNPSRQEDEESTGLGQPLPGAPQPQALSTAGEEEGDKPESKQVVLIDSSY
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining