Learn More
Invitrogen™ Human OLFML2B (aa 302-375) Control Fragment Recombinant Protein
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
Description
Highest antigen sequence indentity to the following orthologs: Mouse (69%), Rat (69%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-66528 (PA5-66528. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.
This gene encodes an olfactomedin domain-containing protein. Most olfactomedin domain-containing proteins are secreted glycoproteins.
Specifications
Specifications
| Accession Number | Q68BL8 |
| Concentration | ≥5.0 mg/mL |
| For Use With (Application) | Blocking Assay, Control |
| Formulation | 1 M urea, PBS with no preservative; pH 7.4 |
| Gene ID (Entrez) | 25903 |
| Name | Human OLFML2B (aa 302-375) Control Fragment |
| Quantity | 100 μL |
| Regulatory Status | RUO |
| Gene Alias | 1110018N05Rik; 4832415H08Rik; AI467542; olfactomedin like 2 B; olfactomedin-like 2 B; olfactomedin-like protein 2 B; Olfml2b; Photomedin-2 |
| Common Name | OLFML2B |
| Show More |