Learn More
Invitrogen™ Human KIF15 (aa 765-858) Control Fragment Recombinant Protein
Product Code. 30211283
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
Product Code. 30211283
Brand: Invitrogen™ RP96411
Recombinant Protein
Highest antigen sequence indentity to the following orthologs: Mouse (76%), Rat (76%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-57304 (PA5-57304. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.
Plus-end directed kinesin-like motor enzyme involved in mitotic spindle assembly. Target protein's mouse homolog associates with microtubulew and appears to play a role in mitosis in fibroblasts and in migration in developing neurons.
TRUSTED_SUSTAINABILITY
Specifications
Accession Number Q9NS87
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 56992
Name Human KIF15 (aa 765-858) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias 3110023M17Rik; 3930402I10Rik; b2b1117.1 aClo; b2b1117.1 Clo; D330038N01; hKLP2; KIF15; kinesin 15; kinesin family member 15; kinesin-like 7; kinesin-like protein 2; Kinesin-like protein 7; kinesin-like protein KIF15; Klp2; Knsl7; N-10 kinesin; NY-BR-62; serologically defined breast cancer antigen NY-BR-62
Common Name KIF15
Gene Symbol KIF15
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence RIDWTKQQEELLSQLNVLEKQLQETQTKNDFLKSEVHDLRVVLHSADKELSSVKLEYSSFKTNQEKEFNKLSERHMHVQLQLDNLRLENEKLLE
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining