Learn More
Invitrogen™ Human PHF15 Control Fragment Recombinant Protein
Product Code. 30210405
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
Product Code. 30210405
Brand: Invitrogen™ RP93570
Recombinant Protein
Highest antigen sequence indentity to the following orthologs: Mouse (80%), Rat (80%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-55275 (PA5-55275. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.
Scaffold subunit of some HBO1 complexes, which have a histone H4 acetyltransferase activity (PubMed:16387653). Acts as a E3 ubiquitin-protein ligase mediating the ubiquitination and subsequent proteasomal degradation of target protein histone demethylase KDM1A (PubMed:25018020). Also acts as a ubiquitin ligase E3 toward itself. Positive regulator of neurogenesis. [UniProt]
TRUSTED_SUSTAINABILITY
Specifications
Accession Number Q9NQC1
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 23338
Name Human PHF15 Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias 1200017K05Rik; AI480685; E3 ubiquitin-protein ligase Jade-2; jade family PHD finger 2; jade family PHD finger protein 2; JADE2; JADE-2; Kiaa0239; mKIAA0239; PHD finger protein 15; Phf15; protein Jade-2; protein Jade-2; E3 ubiquitin-protein ligase Jade-2; si:ch73-202g8.3
Common Name PHF15
Gene Symbol JADE2
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence ASVAADSDVQVLGPAASPKPLGRLRPPRESKVTRRLPGARPDAGMGPPSAVAERPKVSLHFDTETDGYFSDGEMSDSDVEAEDGGVQRGPREAGAEEVVRMGVLA
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining