Learn More
Invitrogen™ Human FOXL2NB (aa 133-170) Control Fragment Recombinant Protein
Product Code. 30207705
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
Product Code. 30207705
Brand: Invitrogen™ RP107836
Recombinant Protein
Highest antigen sequence indentity to the following orthologs: Mouse (33%), Rat (33%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-67199 (PA5-67199. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.
FOXL2NB is a protein coding gene.
TRUSTED_SUSTAINABILITY
Specifications
Accession Number Q6ZUU3
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 401089
Name Human FOXL2NB (aa 133-170) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias C3orf72; FLJ43329 protein; FOXL2 neighbor; FOXL2 neighbor protein; FOXL2NB
Common Name FOXL2NB
Gene Symbol FOXL2NB
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence AGPRNTRRLPAPERERIELAATLCLEGWPLRCLASKGK
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining