Learn More
Invitrogen™ Human SH2D1B (aa 3-30) Control Fragment Recombinant Protein
Product Code. 30206266
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
Product Code. 30206266
Brand: Invitrogen™ RP101476
Recombinant Protein
Highest antigen sequence indentity to the following orthologs: Mouse (79%), Rat (79%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-63020 (PA5-63020. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.
SH2D1B plays a role in controlling signal transduction through at least four receptors, CD84, CD150, CD229 and CD244, expressed on the surface of professional antigen-presenting cells.
TRUSTED_SUSTAINABILITY
Specifications
Accession Number O14796
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 117157
Name Human SH2D1B (aa 3-30) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias Eat2; EAT-2; Eat2a; EAT-2 A; EWS/FLI1 activated transcript 2; EWS/FLI1-activated transcript 2; RGD1563935; SH2 domain containing 1 B; SH2 domain containing 1B1; SH2 domain protein 1B1; SH2 domain-containing molecule EAT2; SH2 domain-containing protein 1 B; Sh2d1b; Sh2d1b1
Common Name SH2D1B
Gene Symbol Sh2d1b
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence LPYYHGRLTKQDCETLLLKEGVDGNFLL
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining