Learn More
Invitrogen™ Human ANKRD22 Control Fragment Recombinant Protein
Product Code. 30205800
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
Product Code. 30205800
missing translation for 'mfr': Invitrogen™ RP90364
Recombinant Protein
Highest antigen sequence indentity to the following orthologs: Mouse (84%), Rat (84%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-53010 (PA5-53010. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.
ANKRD22 is a protein coding gene.
TRUSTED_SUSTAINABILITY
Specifications
Accession Number Q5VYY1
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 118932
Name Human ANKRD22 Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias 5430429D21Rik; Ankrd22; ankyrin repeat domain 22; ankyrin repeat domain-containing protein 22; D19Ertd675e
Common Name ANKRD22
Gene Symbol ANKRD22
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence KTKQNEALVRMLLDAGVEVNATDCYGCTALHYACEMKNPSLIPLLLEARADPTIKNKHGESSLDIARRLKFSQIELMLRK
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining