Learn More
Invitrogen™ Human DEFB106A (aa 22-65) Control Fragment Recombinant Protein
Description
Highest antigen sequence indentity to the following orthologs: Mouse (61%), Rat (61%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-61041 (PA5-61041. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.
Specifications
Specifications
| Accession Number | Q8N104 |
| Concentration | ≥5.0 mg/mL |
| For Use With (Application) | Blocking Assay, Control |
| Formulation | 1 M urea, PBS with no preservative; pH 7.4 |
| Gene ID (Entrez) | 245909 |
| Name | Human DEFB106A (aa 22-65) Control Fragment |
| Quantity | 100 μL |
| Regulatory Status | RUO |
| Gene Alias | BD6; BD-6; Beta-defensin 106; beta-defensin 6; DEFB106; DEFB106A; DEFB106B; DEFB6; DEFB-6; defensin beta 106 A; Defensin, beta 106; defensin, beta 106 A; defensin, beta 6 |
| Common Name | DEFB106A |
| Show More |