Learn More
Invitrogen™ Human DC8 (aa 131-199) Control Fragment Recombinant Protein
Product Code. 30204433
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
Product Code. 30204433
Brand: Invitrogen™ RP104974
Recombinant Protein
Highest antigen sequence indentity to the following orthologs: Mouse (75%), Rat (75%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-65817 (PA5-65817. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.
Part of the MIS12 complex which is required for normal chromosome alignment and segregation and kinetochore formation during mitosis.
TRUSTED_SUSTAINABILITY
Specifications
Accession Number Q96IY1
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 25936
Name Human DC8 (aa 131-199) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias 4833432M17Rik; AI451597; C1orf48; DC31; DC8; Gm105; Gm984; kinetochore-associated protein NSL1 homolog; MIS14; NSL1; NSL1, MIND kinetochore complex component, homolog; NSL1, MIND kinetochore complex component, homolog (S. cerevisiae); NSL1, MIS12 kinetochore complex component; RGD1562052
Common Name DC8
Gene Symbol NSL1
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence RKILECVIKTIKAKQEILKQYHPVVHPLDLKYDPDPAPHMENLKCRGETVAKEISEAMKSLPALIEQGE
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining