Learn More
Invitrogen™ Human ERVMER34-1 (aa 302-442) Control Fragment Recombinant Protein
Description
Highest antigen sequence indentity to the following orthologs: Mouse (23%), Rat (23%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-52836 (PA5-52836. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.
Specifications
Specifications
| Accession Number | Q9H9K5 |
| Concentration | ≥5.0 mg/mL |
| For Use With (Application) | Blocking Assay, Control |
| Formulation | 1 M urea, PBS with no preservative; pH 7.4 |
| Gene ID (Entrez) | 100288413 |
| Name | Human ERVMER34-1 (aa 302-442) Control Fragment |
| Quantity | 100 μL |
| Regulatory Status | RUO |
| Gene Alias | Endogenous retroviral envelope protein HEMO; Endogenous retroviral envelope protein HEMO, 48 kDa form; Endogenous retroviral envelope protein HEMO, secreted form; endogenous retrovirus group MER34 member 1; endogenous retrovirus group MER34 member 1 Env polyprotein; endogenous retrovirus group MER34, member 1; ERVMER34-1; HEMO; HERV-MER_4q12 provirus ancestral Env polyprotein; Human endogenous MER34 (medium-reiteration-frequency-family-34) open reading frame; Human endogenous MER34 ORF; LP9056; LP9056 protein |
| Common Name | ERVMER34-1 |
| Show More |