Learn More
Invitrogen™ Human COL27A1 (aa 1756-1860) Control Fragment Recombinant Protein
Product Code. 30204327
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
Product Code. 30204327
Brand: Invitrogen™ RP93113
Recombinant Protein
Highest antigen sequence indentity to the following orthologs: Mouse (90%), Rat (90%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-61746 (PA5-61746. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.
This gene encodes a member of the fibrillar collagen family, and plays a role during the calcification of cartilage and the transition of cartilage to bone. The encoded protein product is a preproprotein. It includes an N-terminal signal peptide, which is followed by an N-terminal propetide, mature peptide and a C-terminal propeptide. The N-terminal propeptide contains thrombospondin N-terminal-like and laminin G-like domains. The mature peptide is a major triple-helical region. The C-terminal propeptide, also known as COLFI domain, plays crucial roles in tissue growth and repair. Mutations in this gene cause Steel syndrome. Alternatively spliced transcript variants have been found, but the full-length nature of some variants has not been determined.
TRUSTED_SUSTAINABILITY
Specifications
Accession Number Q8IZC6
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 85301
Name Human COL27A1 (aa 1756-1860) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias 5730512J02Rik; AI449266; Col27a1; Collagen alpha-1(XXVII) chain; collagen type XXVII alpha 1; collagen type XXVII alpha 1 chain; collagen type XXVII proalpha 1 chain; collagen, type XXVII, alpha 1; KIAA1870; LOW QUALITY PROTEIN: collagen alpha-1(XXVII) chain; mKIAA1870; procollagen, type XXVII, alpha 1; STLS
Common Name COL27A1
Gene Symbol COL27A1
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence SSEVTQHITIHCLNMTVWQEGTGQTPAKQAVRFRAWNGQIFEAGGQFRPEVSMDGCKVQDGRWHQTLFTFRTQDPQQLPIISVDNLPPASSGKQYRLEVGPACFL
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining