Learn More
Invitrogen™ Human GCSAML (aa 68-133) Control Fragment Recombinant Protein
Product Code. 30201699
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
Product Code. 30201699
Brand: Invitrogen™ RP105777
Recombinant Protein
Highest antigen sequence indentity to the following orthologs: Mouse (29%), Rat (29%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-65012 (PA5-65012. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.
This gene encodes a protein thought to be a signaling molecule associated with germinal centers, the sites of proliferation and differentiation of mature B lymphocytes. Alternate splicing results in multiple transcript variants.
TRUSTED_SUSTAINABILITY
Specifications
Accession Number Q5JQS6
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 148823
Name Human GCSAML (aa 68-133) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias C1orf150; GCSAML; germinal center associated signaling and motility like; germinal center-associated signaling and motility-like protein; germinal center-associated, signaling and motility-like
Common Name GCSAML
Gene Symbol GCSAML
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence CYTVINHIPHQRSSLSSNDDGYENIDSLTRKVRQFRERSETEYALLRTSVSRPCSCTHEHDYEVVF
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining