Learn More
Invitrogen™ Human SOWAHB (aa 515-608) Control Fragment Recombinant Protein
Product Code. 30201623
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30201623 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30201623 Supplier Invitrogen™ Supplier No. RP96746
Recombinant Protein
Highest antigen sequence indentity to the following orthologs: Mouse (70%), Rat (70%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-57828 (PA5-57828. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.
TRUSTED_SUSTAINABILITY
Specifications
Accession Number A6NEL2
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 345079
Name Human SOWAHB (aa 515-608) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias ANKRD56; ankyrin repeat domain 56; ankyrin repeat domain-containing protein 56; Ankyrin repeat domain-containing protein SOWAHB; protein sosondowah homolog B; sosondowah ankyrin repeat domain family member B; SOWAHB
Common Name SOWAHB
Gene Symbol SOWAHB
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence LDEGLLKRSRRPPRSRKPSKAGTAPSPRVDAGLSLKLAEVKAVVAERGWRHSLWVPSGEGSAALAPHRTSEHKSSLVPLDAREHEWIVKLASGS
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining