Learn More
Invitrogen™ Human PAFAH1B2 (aa 123-177) Control Fragment Recombinant Protein
Product Code. 30197089
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
Product Code. 30197089
Brand: Invitrogen™ RP101455
Recombinant Protein
Highest antigen sequence indentity to the following orthologs: Mouse (98%), Rat (98%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-62494 (PA5-62494. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.
Platelet-activating factor (PAF) is a biologically active phospholipid with diverse biologic effects. PAF is degraded to inactive products by hydrolysis of the acetyl group at the sn-2 position to produce the biologically inactive products LYSO-PAF and acetate. This reaction is catalyzed by PAF acetylhydrolase (PAFAH). The various monomeric and multimeric forms of the enzyme are composed of alpha, beta, and gamma PAFAH subunits.
TRUSTED_SUSTAINABILITY
Specifications
Accession Number P68402
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 5049
Name Human PAFAH1B2 (aa 123-177) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias AI747451; AU021353; epididymis secretory protein Li 303; HEL-S-303; intracellular platelet-activating factor acetylhydrolase alpha 2 subunit; mus[b]; PAF acetylhydrolase 30 kDa subunit; PAF-AH 30 kDa subunit; PAF-AH alpha 2; PAFAH subunit beta; PAF-AH subunit beta; PAF-AH1b alpha 2 subunit; Pafah1b2; Pafahb; pl; platelet activating factor acetylhydrolase 1 b catalytic subunit 2; platelet-activating factor acetylhydrolase 1 b, catalytic subunit 2; platelet-activating factor acetylhydrolase 1 b, catalytic subunit 2 (30 kDa); Platelet-activating factor acetylhydrolase alpha 2 subunit; platelet-activating factor acetylhydrolase alpha 2 subunit (PAF-AH alpha 2); platelet-activating factor acetylhydrolase IB subunit beta; platelet-activating factor acetylhydrolase, isoform 1 b, alpha2 subunit; platelet-activating factor acetylhydrolase, isoform 1 b, subunit 2; platelet-activating factor acetylhydrolase, isoform Ib, beta subunit; platelet-activating factor acetylhydrolase, isoform Ib, subunit 2; platelet-activating factor acetylhydrolase, isoform Ib, subunit 2 (30 kDa)
Common Name PAFAH1B2
Gene Symbol PAFAH1B2
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence QLINTRQPQAKIIVLGLLPRGEKPNPLRQKNAKVNQLLKVSLPKLANVQLLDTDG
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining