Learn More
Invitrogen™ Human C1orf131 (aa 192-282) Control Fragment Recombinant Protein
Product Code. 30195933
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
Product Code. 30195933
Brand: Invitrogen™ RP94300
Recombinant Protein
Highest antigen sequence indentity to the following orthologs: Mouse (81%), Rat (81%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-56297 (PA5-56297. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.
The function of this protein remains unknown.
TRUSTED_SUSTAINABILITY
Specifications
Accession Number Q8NDD1
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 128061
Name Human C1orf131 (aa 192-282) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias C1orf131; chromosome 1 open reading frame 131; cPERP-A; Uncharacterized protein C1orf131
Common Name C1orf131
Gene Symbol C1orf131
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence EQERAIMLGAKPPKKSYVNYKVLQEQIKEKKAAKEEEKRLAQETDIFKKKKRKGQEDRKSKKKSAPSILSNGRIGQVGKFKNGTLILSPVD
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining