Learn More
Invitrogen™ Human LRRN4CL (aa 26-106) Control Fragment Recombinant Protein
Product Code. 30182218
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
Product Code. 30182218
Brand: Invitrogen™ RP99930
Recombinant Protein
Highest antigen sequence indentity to the following orthologs: Mouse (67%), Rat (67%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-62168 (PA5-62168. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.
TRUSTED_SUSTAINABILITY
Specifications
Accession Number Q8ND94
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 221091
Name Human LRRN4CL (aa 26-106) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias LRRN4 C-terminal like; LRRN4 C-terminal-like protein; LRRN4CL; UNQ728/PRO1410
Common Name LRRN4CL
Gene Symbol LRRN4CL
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence QDFEEEEADETETAWPPLPAVPCDYDHCRHLQVPCKELQRVGPAACLCPGLSSPAQPPDPPRMGEVRIAAEEGRAVVHWCA
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining