Learn More
CPS1 Antibody (CPS1/8420) [Janelia Fluor™ 525], Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals (Bio-Techne) NBP3-23990JF525
Product Code. 30161009
  • £491.00 / 0.10mL
Description
CPS1 Monoclonal antibody specifically detects CPS1 in Human samples. It is validated for Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY
Specifications
Specifications
CPS1
Monoclonal
Janelia Fluor 525
carbamoyl-phosphate synthase [ammonia], mitochondrial, carbamoyl-phosphate synthase 1, mitochondrial, carbamoyl-phosphate synthetase 1, mitochondrial, carbamoylphosphate synthetase I, Carbamoyl-phosphate synthetase I, CPSase I, CPSASE1, EC 6.3.4.16
Recombinant human CPS1 protein
0.1 mL
Amino Acids Drugs and other small molecules, Cancer, Endocrinology, Signal Transduction
1373
Store at 4°C in the dark.
IgG1 κ
Immunohistochemistry (Paraffin)
CPS1/8420
50mM Sodium Borate
Mouse
Protein A or G purified