Learn More
OXGR1/GPR80/P2Y15 Antibody (JE64-04), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-32684
This item is not returnable.
View return policy
Beschreibung
OXGR1/GPR80/P2Y15 Monoclonal antibody specifically detects OXGR1/GPR80/P2Y15 in Human,Mouse,Rat samples. It is validated for Western Blot
Spezifikation
| OXGR1/GPR80/P2Y15 | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:500-1:2000 | |
| Alpha-ketoglutarate receptor 1, G protein-coupled receptor 80, G protein-coupled receptor 99, GPR802-oxoglutarate receptor 1, GPR99, G-protein coupled receptor 80, G-protein coupled receptor 99, MGC119206, MGC119207, MGC119208, oxoglutarate (alpha-ketoglutarate) receptor 1, P2RY15P2Y-like GPCR, P2Y purinoceptor 15, P2Y15P2Y-like nucleotide receptor, seven transmembrane helix receptor | |
| Synthetic peptide within human OXGR1/GPR80/P2Y15 aa 171-220/337. (Uniprot: Q96P68) | |
| 100 μL | |
| GPCR | |
| 27199 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| JE64-04 | |
| Unconjugated | |
| 1*TBS (pH7.4), 0.05% BSA and 40% Glycerol | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur