Learn More
TMEFF2/Tomoregulin-2 Antibody (Janssen patent anti-TMEFF2), Alexa Fluor™ 405, Novus Biologicals™
Human Monoclonal Antibody
Brand:  Novus Biologicals NBP3-27983AF405
Product Code. 30114403
  • £328.00 / 0.10mL
Description
TMEFF2/Tomoregulin-2 Monoclonal antibody specifically detects TMEFF2/Tomoregulin-2 in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
TRUSTED_SUSTAINABILITY
Specifications
Specifications
TMEFF2/Tomoregulin-2
Monoclonal
Alexa Fluor 405
CT120.2, HPP1member 2, Hyperplastic polyposis protein 1, TPEFtomoregulin, TR-2, transmembrane protein TENB2, Transmembrane protein with EGF-like and two follistatin-like domains, transmembrane protein with EGF-like and two follistatin-like domains 2, TRtomoregulin-2
TMEFF2
0.1 mL
Cancer, Cell Biology, Cytokines and Growth Factors
23671
Store at 4°C in the dark.
IgG1
Flow Cytometry, ELISA, Functional Assay
Janssen patent anti-TMEFF2
50mM Sodium Borate
Human
Protein A purified