Learn More
Novus Biologicals™ ZSCAN5 Antibody (PCRP-ZSCAN5A-2H4) [mFluor Violet 610 SE], Novus Biologicals™
Product Code. 30066025 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30066025 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30066025 Supplier Novus Biologicals™ Supplier No. NBP327033MFV610
Mouse Monoclonal Antibody
ZSCAN5 Monoclonal antibody specifically detects ZSCAN5 in Human samples. It is validated for Flow Cytometry,Immunocytochemistry
TRUSTED_SUSTAINABILITY
Specifications
Antigen ZSCAN5
Applications Flow Cytometry, Immunocytochemistry
Classification Monoclonal
Clone PCRP-ZSCAN5A-2H4
Conjugate mFluor Violet 610 SE
Formulation 50 mM Sodium Borate
Gene Alias MGC4161, zinc finger and SCAN domain containing 5, zinc finger and SCAN domain containing 5A, zinc finger and SCAN domain-containing protein 5A, Zinc finger protein 495ZSCAN5, ZNF495
Host Species Mouse
Immunogen Recombinant fragment (around aa37-124) of human ZSCAN5 protein
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 79149
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2b