Learn More
Value Plastics Sanitary to 600 Series Hose Barb Fittings
Product Code. 30053642 Shop All Cole-Parmer Products
Change view
Click to view available options
Diameter (English) Inner:
1 in.
1/2 in.
1/4 in.
3/4 in.
3/8 in.
Includes:
1-1/2 in. Sanitary Clamp Size
3/4 in. Sanitary Clamp Size
Unit Size:
Pack of 100
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (English) Inner Includes unitSize
30053642 1 in. 1-1/2 in. Sanitary Clamp Size Pack of 100
30053491 1/2 in. 3/4 in. Sanitary Clamp Size Pack of 100
30054350 3/4 in. 3/4 in. Sanitary Clamp Size Pack of 100
30053971 3/8 in. 1-1/2 in. Sanitary Clamp Size Pack of 100
30054233 3/4 in. 1-1/2 in. Sanitary Clamp Size Pack of 100
30054212 1/4 in. 1-1/2 in. Sanitary Clamp Size Pack of 100
30053786 3/8 in. 3/4 in. Sanitary Clamp Size Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Product Code. 30053642 Supplier Value Plastics Supplier No. 5011869
Only null left
Add to basket
Add to basket
Sanitary fittings provide a secure connection where preventing contamination is crucial
Reduce the risk of bacterial growth with smooth surfaces and minimal crevices. Easily transition from sanitary to hose barb connection. Maintain a low profile and ease of assembly with 600 series barb fittings
Specifications
Certifications/Compliance ADCF;FDA;Non-DEHP;Non-Phthalate;RoHS;USP
Diameter (English) Inner 1 in.
Packaging Quantity 100/Pk.
Outlet Port Sanitary x 600 Series Barb
Sterilization Method Gamma radiation, ethylene oxide (EtO), E-beam, and autoclave
Shape Straight
Includes 1-1/2 in. Sanitary Clamp Size