Learn More
Sartorius Cubis II MTS Balance
Shop All Sartorius Products
Click to view available options
Capacity:
120 g
8200 g
Readability:
0.01
10 mg
Description:
MCA Display including touch, scroll, swipe functionality with factory-installed essential weighing applications and diverse QPapp Packages for optional software extensions
MCE Display with factory-installed essential weighing applications
Unit Size:
Each
Description
- Leading performance.
- Monolithical weighing system, integrated climate sensors, individual sample holders
- Error-free operation.
- Individual QApp workflows, motorized auto-leveling
- The modular weighing system Cubis™ II can be individually configured from different components.
Specifications
Specifications
| Connection Type | D-sub connector, 9-pin |
| Description | MCA Display including touch, scroll, swipe functionality with factory-installed essential weighing applications and diverse QPapp Packages for optional software extensions |
| Capacity | 8200 g |
| Display Type | Color Touchscreen |
| Readability | 10 mg |
| Material | Die-cast Aluminum, Plastic, Optiwhite Float Glass and Stainless Steel |
| Model | Cubis II MTS Balance |
| Stabilization Time | 1 second |
| Type | Balance |
| Electrical Requirements | 00 to 240 VAC (±10%) |
| Show More |