Learn More
Desvenlafaxine Succinate Monohydrate (O-Desmethylvenlafaxine Succinate Monohydrate), Mikromol™
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
Description
Desvenlafaxine Succinate Monohydrate (O-Desmethylvenlafaxine Succinate Monohydrate)
Specifications
Specifications
| Chemical Name or Material | Desvenlafaxine Succinate Monohydrate |
| CAS | 386750-22-7 |
| Analyte or Component Names | Desvenlafaxine Succinate Monohydrate |
| Shipping Condition | Ice pack (-20°C) |
| Molecular Formula | C16 H25 N O2 . C4 H6 O4 . H2 O |
| Certifications/Compliance | ISO 17025 |
| Synonym | Butanedioic acid, compd. with 4-[2-(dimethylamino)-1-(1-hydroxycyclohexyl)ethyl]phenol, hydrate (1:1:1), Butanedioic acid, compd. with 4-[2-(dimethylamino)-1-(1-hydroxycyclohexyl)ethyl]phenol (1:1), monohydrate (9CI) |
| InChI Formula | InChI=1S/C16H25NO2.C4H6O4.H2O/c1-17(2)12-15(13-6-8-14(18)9-7-13)16(19)10-4-3-5-11-16;5-3(6)1-2-4(7)8;/h6-9,15,18-19H,3-5,10-12H2,1-2H3;1-2H2,(H,5,6)(H,7,8);1H2 |
| SMILES | O.CN(C)CC(c1ccc(O)cc1)C2(O)CCCCC2.OC(=O)CCC(=O)O |
| IUPAC Name | butanedioic acid;4-[2-(dimethylamino)-1-(1-hydroxycyclohexyl)ethyl]phenol;hydrate |
| Show More |