Learn More
TRAF-2 Antibody (9A5), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand:  Novus Biologicals NBP3-26271-100ul
 View more versions of this product
Product Code. 30037375
  • £406.00 / 100µL
Description
TRAF-2 Monoclonal antibody specifically detects TRAF-2 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunoprecipitation
TRUSTED_SUSTAINABILITY
Specifications
Specifications
TRAF-2
Monoclonal
Unconjugated
PBS, pH 7.4, 150mM NaCl and 50% glycerol
Rabbit
Affinity purified
RUO
Primary
Human
Purified
Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunoprecipitation
9A5
Western Blot 1:500-1:5000, Flow Cytometry 1:20-1:200, ELISA, Immunohistochemistry 1:50-1:200, Immunoprecipitation 1:200-1:1000
E3 ubiquitin-protein ligase TRAF2, EC 6.3.2.-, MGC:45012, TNF receptor-associated factor 2, TRAP3TRAP, tumor necrosis factor type 2 receptor associated protein 3, Tumor necrosis factor type 2 receptor-associated protein 3
A synthesized peptide derived from Human TRAF-2 [UniProt Q12933]
100 μL
Apoptosis, Cancer, Cytokine Research, Zinc Finger
7186
Store at -20 to -70C. Avoid freeze-thaw cycles.