Description
XPB Polyclonal antibody specifically detects XPB in Human,Mouse,Rabbit,Virus samples. It is validated for Immunoprecipitation,Western Blot,ChIP assay (ChIP)
Specifications
Specifications
| Antigen | XPB |
| Applications | Immunoprecipitation, Western Blot, ChIP Assay |
| Classification | Polyclonal |
| Conjugate | APC |
| Formulation | PBS |
| Gene Alias | Basic transcription factor 2 89 kDa subunit, BTF2, BTF2 p89, DNA excision repair protein ERCC-3, DNA repair protein complementing XP-B cells, EC 3.6.1, EC 3.6.4.12, excision repair cross-complementing rodent repair deficiency, complementationgroup 3 (xeroderma pigmentosum group B complementing), GTF2H, RAD25, TFIIH, TFIIH basal transcription factor complex 89 kDa subunit, TFIIH basal transcription factor complex helicase XPB subunit, TFIIH p89, Xeroderma pigmentosum group B-complementing protein, xeroderma pigmentosum, complementation group B, XPBC, XPBTFIIH 89 kDa subunit |
| Host Species | Rabbit |
| Immunogen | The immunogen recognized by this antibody maps to a region between residue 732 and 782 of human excision repair cross-complementing rodent repair deficiency, complementation group 3 using the numbering given in entry NP_000113.1 (GeneID 2071). |
| Purification Method | Immunogen affinity purified |
| Quantity | 0.1 mL |
| Show More |