Learn More
R&D Systems™ Human Total E-Cadherin DuoSet IC ELISA
Product Code. 30021550
Click to view available options
Packaging Quantity:
15 Plates
2 Plates
5 Plates
Unit Size:
1 set
Product Code. 30021550
Brand: R&D Systems™ DYC42252
Contains the basic components required for the development of sandwich ELISAs to measure in cell lysates.
The Human Total E-Cadherin DuoSet IC ELISA is a Solid Phase Sandwich ELISA that quantifies human E-Cadherin.
TRUSTED_SUSTAINABILITY
Specifications
Product Type ELISA Development Kit for Intracellular Proteins
Quantity 1 Kit
Species Human
Conjugate Biotin
Assay Range 156.00 to 10000 pg/mL
Cross Reactivity <50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules
For Use With (Application) ELISA
Gene ID (Entrez) 999
Packaging Quantity 2 Plates
Content And Storage Store the unopened product at 2°C to 8°C. Do not use past expiration date.