Learn More
Melford PMSF
Click to view available options
Quantity:
5 g
10 g
25 g
Unit Size:
10g
25g
5g
Description
Irreversible serine protease inhibitor of chymotrypsin, kallikrein, papain, subtilisin, thrombin and trypsin.
Irreversible serine protease inhibitor of chymotrypsin, kallikrein, papain, subtilisin, thrombin and trypsin. Working Conentration: 0.1-1.0mM.
Specifications
Specifications
| CAS | 329-98-6 |
| Molecular Formula | C7H7FO2S |
| Quantity | 25 g |