Learn More
Human Caldesmon/CALD1 Antibody, R&D Systems™
Mouse Monoclonal Antibody
Brand: R&D Systems MAB7569-SP
This item is not returnable.
View return policy
Description
Caldesmon/CALD1 Monoclonal specifically detects Caldesmon/CALD1 in Human samples. It is validated for Western Blot, Immunocytochemistry.
Specifications
| Caldesmon/CALD1 | |
| Monoclonal | |
| LYOPH | |
| Western Blot 2 ug/mL, Immunocytochemistry 8-25 ug/mL | |
| Q05682 | |
| CALD1 | |
| E. coli-derived recombinant human Caldesmon/CALD1 Lys696-Val793 Accession # Q05682 | |
| 25 μg | |
| Primary | |
| Detects human Caldesmon/CALD1 in direct ELISAs and Western blots. | |
| Human | |
| Purified |
| Western Blot, Immunocytochemistry | |
| 767001 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| CAD, CALD1, caldesmon, caldesmon 1, CDM, CDMH-CAD, HCAD, LCAD, L-CAD, MGC21352, NAG22 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 800 | |
| Sterile PBS to a final concentration of 0.5 mg/mL. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2b |