Learn More
R&D Systems™ Cynomolgus Monkey CCL3/MIP-1 alpha (Unique) VersaClone cDNA
Click to view available options
Quantity:
10 μg
Unit Size:
10µg
Description
- CCL3 is a small inducible cytokine.
- It plays a role in inflammatory responses through binding to the receptors CCR1, CCR4 and CCR5.
- CCL3 expression can be induced in a variety of hematopoietic cells, fibroblasts, smooth muscle cells, and epithelial cells.
- Polymorphisms at this locus may be associated with both resistance and susceptibility to infection by human immunodeficiency virus type 1.
- The insert is sequence verified and the entire plasmid DNA and insert translated sequences are provided.
- For reasearch use only. Not for use in humans.
Specifications
Specifications
| Gene | cynoCCL3 |
| Accession Number | unique |
| Concentration | 10 μg at 0.2 μg/μL |
| Formulation | cDNA is provided in 10 mM Tris-Cl, pH 8.5 |
| Insert Type | 295 bp |
| Gene ID (Entrez) | 6348 |
| Content And Storage | Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. |
| Species | Cynomolgus Monkey |
| Quantity | 10 μg |