Learn More
Human/Mouse Renin R Antibody, R&D Systems™
Goat Polyclonal Antibody
Brand: R&D Systems AF5716-SP
This item is not returnable.
View return policy
Description
Renin R Polyclonal specifically detects Renin R in Human, Mouse samples. It is validated for Western Blot.
Specifications
| Renin R | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| APT6M8-9, ATP6AP2, ATP6IP2, ATP6M8-9, ATP6M8-9MRXE, ATPase H(+)-transporting lysosomal accessory protein 2, ATPase H(+)-transporting lysosomal-interacting protein 2, ATPase, H+ transporting, lysosomal accessory protein 2, ATPase, H+ transporting, lysosomal interacting protein 2, CAPER, ELDF10, Embryonic liver differentiation factor 10, ER-localized type I transmembrane adaptor, H+ transporting, lysosomal (vacuolar proton pump) membrane sectorassociated protein M8-9, HT028, M8-9, MGC99577, MRXE, MSTP009, N14F, renin receptor, ReninR, vacuolar proton ATP synthase membrane sector associated protein M8-9, V-ATPase M8.9 subunit, XMRE | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 10159 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot | |
| LYOPH | |
| Western Blot 1 ug/mL | |
| O75787 | |
| ATP6AP2 | |
| E. coli-derived recombinant human Renin R Asn17-Glu302 Accession # O75787 | |
| 25 μg | |
| Primary | |
| Detects human and mouse Renin R in Western blots. | |
| Human, Mouse | |
| IgG |