Learn More
Mouse NELL2 Antibody, R&D Systems™
Sheep Polyclonal Antibody
Brand: R&D Systems AF6736-SP
This item is not returnable.
View return policy
Description
NELL2 Polyclonal specifically detects NELL2 in Mouse samples. It is validated for Immunohistochemistry.
Specifications
| NELL2 | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| Mel91, nel (chicken)-like 2, NEL-like 2 (chicken), NEL-like protein 2, Nel-related protein 2, NRP2, NRP2neural epidermal growth factor-like 2, protein kinase C-binding protein NELL2 | |
| Sheep | |
| Affinity Purified | |
| RUO | |
| 4753 | |
| Sterile PBS to a final concentration of 0.2 mg/mL. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Immunohistochemistry | |
| LYOPH | |
| Immunohistochemistry 5-15 ug/mL | |
| NP_058023 | |
| NELL2 | |
| Mouse myeloma cell line NS0-derived recombinant mouse NELL2 Leu25-Leu819 Accession # NP_058023 | |
| 25 μg | |
| Primary | |
| Detects mouse NELL2 in direct ELISAs. In direct ELISAs, approximately 15% cross-reactivity with recombinant human (rh) NELL2 and less than 3% cross-reactivity with rhNELL1 is observed. | |
| Mouse | |
| IgG |