Learn More
Human PLTP Antibody, R&D Systems™
Sheep Polyclonal Antibody
Brand: R&D Systems AF5109-SP
This item is not returnable.
View return policy
Description
PLTP Polyclonal specifically detects PLTP in Human samples. It is validated for Western Blot, Knockout Validated.
Specifications
| PLTP | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| HDLCQ9, Lipid transfer protein II, phospholipid transfer protein | |
| Sheep | |
| Affinity Purified | |
| RUO | |
| 5360 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot, Immunoassay | |
| LYOPH | |
| Western Blot 1 ug/mL, Knockout Validated | |
| P55058 | |
| PLTP | |
| Mouse myeloma cell line NS0-derived recombinant human PLTP isoform 1 Glu18-Val493 (Glu18Val) Accession # P55058 | |
| 25 μg | |
| Primary | |
| Detects human PLTP in direct ELISAs and Western blots. In direct ELISAs, approximately 10% cross-reactivity with recombinant mouse PLTP is observed. | |
| Human | |
| IgG |