Learn More
Human/Mouse SSEA-3 Antibody, R&D Systems™
GREENER_CHOICE
Rat Monoclonal Antibody has been used in 17 publications
Brand:  R&D Systems MAB1434-SP
Product Code. 18795548
  • £170.00 / 25µg
Description
SSEA-3 Monoclonal specifically detects SSEA-3 in Human, Mouse samples. It is validated for Flow Cytometry, Immunocytochemistry, CyTOF-ready.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
SSEA-3
Monoclonal
LYOPH
Flow Cytometry 2.5 ug/10^6 cells, Immunocytochemistry 8-25 ug/mL, CyTOF-ready
SSEA3, Stage specific embryonic antigen 3, Stage-specific embryonic antigen 3
Four to eight cell stage mouse embryos
25 μg
Primary
Reconstitute at 0.5 mg/mL in sterile PBS.
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Flow Cytometry, Immunocytochemistry, CyTOF
MC-631
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Rat
Affinity Purified
RUO
Recognizes a carbohydrate epitope of SSEA-3 (4, 5).
Human, Mouse
IgM