Learn More
Human Vimentin Antibody, R&D Systems™
Rat Monoclonal Antibody
Brand: R&D Systems MAB21051-SP
This item is not returnable.
View return policy
Description
Vimentin Monoclonal specifically detects Vimentin in Human samples. It is validated for Western Blot.
Specifications
| Vimentin | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1 ug/mL | |
| P08670 | |
| VIM | |
| E. coli-derived recombinant human Vimentin Ser2-Glu466 Accession # P08670 | |
| 25 μg | |
| Primary | |
| Detects human Vimentin in direct ELISAs and Western blots. | |
| Human | |
| Purified |
| Western Blot | |
| 280622 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| FLJ36605, VIM, vimentin | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 7431 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2a |