Learn More
Human PLD1 Antibody, R&D Systems™
Goat Polyclonal Antibody
Brand: R&D Systems AF5615-SP
This item is not returnable.
View return policy
Description
PLD1 Polyclonal specifically detects PLD1 in Human samples. It is validated for Western Blot.
Specifications
| PLD1 | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| Choline phosphatase 1, EC 3.1.4.4, hPLD1, Phosphatidylcholine-hydrolyzing phospholipase D1, phospholipase D1, phospholipase D1, phophatidylcholine-specific, phospholipase D1, phosphatidylcholine-specific, PLD 1 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 5337 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot | |
| LYOPH | |
| Western Blot 1 ug/mL | |
| Q13393 | |
| PLD1 | |
| E. coli-derived recombinant human PLD1 Met1-Pro140 Accession # Q13393 | |
| 25 μg | |
| Primary | |
| Detects endogenous human PLD1 in Western blots. | |
| Human | |
| IgG |