Learn More
Human/Mouse/Rat Neurofascin Antibody, R&D Systems™
GREENER_CHOICE
Mouse Monoclonal Antibody
Brand:  R&D Systems MAB3235-SP
Product Code. 18788799
  • £168.00 / 25µg
Description
Neurofascin Monoclonal specifically detects Neurofascin in Human, Mouse, Rat samples. It is validated for Western Blot.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Neurofascin
Monoclonal
LYOPH
Western Blot 0.5 ug/mL
NP_446361
NFASC
Mouse myeloma cell line NS0-derived recombinant rat Neurofascin Ile25-Ala1031 Accession # NP_446361
25 μg
Primary
Detects rat Neurofascin in direct ELISAs and Western blots.
Human, Mouse, Rat
Purified
Western Blot
727030
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
DKFZp686P2250, KIAA0756FLJ46866, neurofascin, neurofascin homolog, neurofascin homolog (chicken), NF, NFASC, NRCAML
Mouse
Protein A or G purified from hybridoma culture supernatant
RUO
23114
Sterile PBS to a final concentration of 0.5 mg/mL.