Learn More
Human CLOCK Antibody, R&D Systems™
Sheep Polyclonal Antibody
Brand: R&D Systems AF4026-SP
This item is not returnable.
View return policy
Description
CLOCK Polyclonal specifically detects CLOCK in Human samples. It is validated for Western Blot.
Specifications
| CLOCK | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| BHLHE8, circadian locomoter output cycles protein kaput, Class E basic helix-loop-helix protein 8, clock (mouse) homolog, clock homolog (mouse), EC 2.3.1.48, hCLOCK, KAT13D, KIAA0334bHLHe8circadian locomoter output cycles kaput protein | |
| Sheep | |
| Affinity Purified | |
| RUO | |
| 9575 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot | |
| LYOPH | |
| Western Blot 0.5 ug/mL | |
| O15516 | |
| CLOCK | |
| E. coli-derived recombinant human CLOCK Glu108-Glu234 Accession # O15516 | |
| 25 μg | |
| Primary | |
| Detects human CLOCK in Western blots. | |
| Human | |
| IgG |