Learn More
Human Protogenin Antibody, R&D Systems™
Sheep Polyclonal Antibody
Brand: R&D Systems AF6795-SP
This item is not returnable.
View return policy
Description
Protogenin Polyclonal specifically detects Protogenin in Human samples. It is validated for Immunohistochemistry.
Specifications
| Protogenin | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| FLJ12795, FLJ13221, FLJ25756, IGDCC5, immunoglobulin superfamily, DCC subclass, member 5, Protein Shen-Dan, protogenin, protogenin homolog (Gallus gallus), PRTG, RPTG, Shen-Dan | |
| Sheep | |
| Affinity Purified | |
| RUO | |
| 283659 | |
| Sterile PBS to a final concentration of 0.2 mg/mL. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Immunohistochemistry | |
| LYOPH | |
| Immunohistochemistry 5-15 ug/mL | |
| Q2VWP7 | |
| PRTG | |
| Mouse myeloma cell line NS0-derived recombinant human Protegenin Phe36-Ala952 Accession # Q2VWP7 | |
| 25 μg | |
| Primary | |
| Detects human Protegenin in direct ELISAs. In direct ELISAs, approximately 30% cross-reactivity with recombinant mouse Protegenin is observed. | |
| Human | |
| IgG |