Learn More
Human Sirtuin 3/SIRT3 Antibody, R&D Systems™
Mouse Monoclonal Antibody
Brand: R&D Systems MAB7488-SP
This item is not returnable.
View return policy
Description
Sirtuin 3/SIRT3 Monoclonal specifically detects Sirtuin 3/SIRT3 in Human samples. It is validated for Western Blot.
Specifications
| Sirtuin 3/SIRT3 | |
| Monoclonal | |
| LYOPH | |
| Western Blot 2 ug/mL | |
| Q9NTG7 | |
| SIRT3 | |
| E. coli-derived recombinant human Sirtuin 3/SIRT3 Ser101-Lys399 Accession # Q9NTG7 | |
| 25 μg | |
| Primary | |
| Detects human Sirtuin 3/SIRT3 in ELISAs. In direct ELISAs, no cross-reactivity with recombinant human Sirtuin 2 is observed. | |
| Human | |
| Purified |
| Western Blot | |
| 850911 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| EC 3.5.1, EC 3.5.1.-, hSIRT3, mitochondrial nicotinamide adenine dinucleotide-dependent deacetylase, NAD-dependent deacetylase sirtuin-3, mitochondrial, SIR2L3, SIR2L3silent mating type information regulation 2, S.cerevisiae, homolog 3, sir2-like 3, SIR2-like protein 3, SIRT3, sirtuin (silent mating type information regulation 2 homolog) 3 (S. cerevisiae), sirtuin (silent mating type information regulation 2, S.cerevisiae, homolog) 3, sirtuin 3, sirtuin type 3 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 23410 | |
| Sterile PBS to a final concentration of 0.5 mg/mL. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG1 |